Thermo Scientific™ Shandon™ Fine Point Dissecting Scissors
Achieve consistent quality when dissecting with Thermo Scientific™ Shandon™ Fine Point Dissection Scissors.
Manufacturer: Thermo Scientific™ 711999
Description
Achieve consistent quality when dissecting with Thermo Scientific™ Shandon™ Fine Point Dissection Scissors. Available in premium and standard grades.
- Properly hardened, tempered and passivated
- Electronically polished and cleaned for a high-quality finish
- Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
- Guaranteed to be free from defects in material and workmanship
Specifications
Premium | |
Fine Point | |
4.5 in. |
11.4cm | |
Fine Point Dissecting Scissors |