missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Epredia™ EZ Single Cytofunnel™ with Brown Filter Cards
Use the Epredia™ EZ Single Cytofunnel™ with Brown Filter Cards for sample volumes of 0.4 mL or less.
Supplier: Epredia™ A78710004
Description
- Cell deposition area of 6 mm in diameter (28 mm2)
- Pre-attached brown filter card for slower absorbency
Specifications
Cytofunnel with brown filter cards and caps | |
40/Cs. | |
Cytofunnel with Brown Filter Card | |
0.4 mL |