Please login to your online account to display your discounted pricing
Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs
Hold tissue specimens during processing and embedding with Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs.
Manufacturer: thermo scientific™  B1000729PK
 View more versions of this product
Catalog No. B1000729PK
  • / Case of 750 Each
Description & Specifications
Specifications
Color Pink
Material Acetal polymer
Writing Face Angle 45°
Pore Style Slotted
Hold tissue specimens during processing and embedding with Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs. The cassettes are compatible with Thermo Scientific™ Shandon™ MicroWriter™ cassette labelers.
  • Acetal polymer cassettes with writing face at 45° angle
  • Slotted pore style
  • Locking lid attached by two hinges
Ordering Information:
  • Tube packs are supplied in 10 tubes of 75 cassettes each