Please login to your online account to display your discounted pricing
Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs
Hold tissue specimens during processing and embedding with Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs.
Manufacturer: thermo scientific™  B1000729AQ
 View more versions of this product
Catalog No. B1000729AQ
  • / Pack of 750 Each
Description & Specifications
Specifications
Pore Style Slotted
Material Acetal polymer
Writing Face Angle 45°
Color Aqua
Hold tissue specimens during processing and embedding with Thermo Scientific™ Shandon™ Embedding Cassettes II in Tube Packs. The cassettes are compatible with Thermo Scientific™ Shandon™ MicroWriter™ cassette labelers.
  • Acetal polymer cassettes with writing face at 45° angle
  • Slotted pore style
  • Locking lid attached by two hinges
Ordering Information:
  • Tube packs are supplied in 10 tubes of 75 cassettes each