Epredia™ Shandon™ EA-65 Stain
Improve the expression of eosin-stained components with Epredia™ Shandon™ EA-65 Stain, which contains a lower concentration of light green.
$295.03 - $295.03
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6766003
|
Epredia™
6766003 |
2 x 1L |
Case of 2 Each for $295.03
|
|
|||||
Description
- Polychromatic cytoplasmic stain
- Ideal for thicker non-GYN preparations
- Biological Stain Commission-certified dye
Specifications
Cellular Components Stain |