Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs
Aid the process of speedy and reliable biopsy specimen labeling with Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs.
Manufacturer:  Thermo Scientific™ B1000730WH
 View more versions of this product
Catalog No. B1000730WH
Specifications
Description
Aid the process of speedy and reliable biopsy specimen labeling with Thermo Scientific™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs. These cassettes are for use with the MicroWriter Cassette Labeler or for convenient storage.
  • Writing face is flat 45° angle
  • Locking lid attached by two hinges
  • Front or back opening
Ordering Information:
  • Packaged in 75-cassette tubes
Description & Specifications
Specifications
Biopsy Cassettes, Small Square Pore
45°
White