Epredia™ Shandon™ Autopsy Headrests, Rubber
Epredia™ Shandon Autopsy Headrests feature beveled corners to protect autopsy surfaces from scratches.
$824.47 - $824.47
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
38-108-43
|
Epredia™
SH407 |
Rubber |
Each for $824.47
|
|
|||||
Description
Features:- Ten positions of height, width, length and curvature allow for maximum accommodation.
- Each side of the unit is deeply curved to hold the head securely, and beveled corners protect surfaces.
- Only four points contact the table and base is curved to permit drainage materials to flow freely.
- Available in autoclavable aluminum or semi-hard rubber
Specifications
Autopsy Headrest |