Please login to your online account to display your discounted pricing

Thermo Scientific™ Shandon™ AAdson Fine Serrated Thumb Forceps

Choose 4.75″ (12.1cm) Shandon™ Adson Thumb Forceps with a fine serrated edge for quality and precision.

$59.71 - $59.71

Specifications

Type Adson
Length (English) 4.75 in.
Length (Metric) 12.1cm
Shape Straight
View More Specs
Products
Catalog Number Mfr. No. Grade Price Quantity    

38-106-61

 
thermo scientific™
SH626
Standard Each for $59.71
Description & Specifications

Specifications

Type Adson
Length (English) 4.75 in.
Length (Metric) 12.1cm
Shape Straight

Choose the 4.75″ (12.1cm) Thermo Scientific™ Shandon™ Adson Thumb Forceps with a fine serrated edge for quality and precision. Forceps are available in premium and standard grades.

Features:

  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards