Thermo Scientific™ Shandon™ 450 Autopsy Saw
Cut bone, not soft tissue with the Thermo Scientific™ Shandon™ 450 Autopsy Saw. Its blade oscillates at high speed.
$1,106.33 - $4,123.31
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Voltage Price Quantity  
Catalog Number Mfr. No. Description Voltage Price Quantity  
38-103-57 Thermo Scientific™
SH450
Saw 110/120V Each for $4,123.31
38-103-64 Thermo Scientific
SH454
Foot Switch 110/120V Each for $1,106.33
Specifications
Description
Cut bone, not soft tissue with the Thermo Scientific™ Shandon™ 450 Autopsy Saw. Its blade oscillates at high speed with a flexible drive shaft that allows for easy guidance of the saw's path.
Features:
  • Safety flange permits firm grip
  • Autoclavable handpiece stays cool during operation, detaches for cleaning and storage
  • Saw attaches to the HEPA Bone Dust Collector (10001, 10022)
Physical Specifications:
  • Electrical Requirements: 110-120V, 50/60Hz (450); 220-240V, 50/60Hz (45022)
  • Handpiece weight: 1.5 lb. (0.7kg)
  • Arbor removes with 0.125in. (0.3cm) Allen Wrench (1075309)
  • Optional foot switch available (454, 45422)
Includes:
  • 8′ (243.8cm) service cord with hospital-grade plug and fixed arbor large section blade (10003)