Please login to your online account to display your discounted pricing

Tocris™ SGS 518 oxalate

Selective 5-HT6 antagonist

$244.81 - $997.23

Specifications

Product Type SGS 518 oxalate
Inhibitors SGS 518 oxalate
Molecular Weight 510.51
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

368810R

 
tocris™
RDS368810
10mg Each for $244.81

368850R

 
tocris™
RDS368850
50mg Each for $997.23
Description & Specifications

Specifications

Product Type SGS 518 oxalate
Inhibitors SGS 518 oxalate
Molecular Weight 510.51
Purity >98

Selective antagonist of the 5-HT6 subtype of the serotonin receptor. Useful for the treatment of cognitive impairment that is associated with schizophrenia and Alzheimer's disease.

  • Chemical Name: 1-Methyl-3-(1-methyl-4-piperindin-4-yl)-5-hydroxy-1H-indole 2,6-difluorobenzenesulfonic acid ester oxalate
  • Formula: C21H22F2N2O3S.C2H2O4
  • CAS Number: 445441-27-0
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >98