Invitrogen™ SGK2 Recombinant Human Protein DFS Item
SGK2 is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK).
Manufacturer:  Invitrogen™ LSPV4057
 View more versions of this product
Catalog No. PV4057
Description
Description
Unlike SGK, SGK2 is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK.
Specifications
Specifications
NP_057360, S416D
Frozen
69.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Catalytic
Kinase Assay
SGK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
SGK2
Baculovirus
Human
GST Tag
Documents