Please login to your online account to display your discounted pricing

SGE™ Varian 1075/1077 Liners: Splitless


Manufacturer: sge™  SG092024

 View more versions of this product

Catalog No. SG092024

  • / Each
 

Description & Specifications

Specifications

Product Type Varian 1075/1077 Splitless Liner with slots at both ends
For Use With (Equipment) SGE Varian 1075/1077 Liners
Diameter (Metric) Inner 2mm
Diameter (Metric) Outer 6.3mm
Length (Metric) 74mm
Quantity 5/Pk.