missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGE™ Varian 1075/1077 Liners: Splitless

$192.56 - $192.56

Specifications

Diameter (Metric) Inner 2mm
Diameter (Metric) Outer 6.3mm
For Use With (Equipment) SGE Varian 1075/1077 Liners
Length (Metric) 74mm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
SG092024 SGE™
SG092024
5/Pk. Each for $192.56
Specifications Description & Specifications

Specifications

2mm
6.3mm
SGE Varian 1075/1077 Liners
74mm