missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGE™ Varian 1075/1077 Liners: Split

$265.09 - $265.09

Specifications

Diameter (Metric) Inner 4mm
Diameter (Metric) Outer 6.3mm
For Use With (Equipment) Varian 1075/1077
Length (Metric) 72mm
Quantity 5/Pk.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
SG092021 SGE™
SG092021
Split liner with quartz wool Each for $265.09
Specifications Description & Specifications

Specifications

4mm
Varian 1075/1077
5/Pk.
6.3mm
72mm