missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGE™ Thermo Scientific™ GC Autosampler Syringes

Incorporate SGE's Diamond Syringe Technology

Manufacturer:  SGE™ SG000655

 View more versions of this product

Catalog No. SG000655


 

Specifications Description & Specifications

Specifications

Varian/Bruker
Removable
2 in.
8mm
Gas chromatography
0.018 in.
Cone
26 ga
0.314 in.
1μL
GC Autosampler Syringe
0.47mm
1μL
Each
51mm
51mm