SGE™ Thermo Scientific™ GC Autosampler Syringes
Incorporate SGE's Diamond Syringe Technology
$254.54 - $254.54
Specifications
Type GC Autosampler Syringe
For Use With (Application) Gas chromatography
Needle Point Style Cone
Quantity Each
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Connection Type Diameter (English) Outer Needle Length (English) Needle Needle Gauge Price Quantity  
Catalog Number Mfr. No. Connection Type Diameter (English) Outer Needle Length (English) Needle Needle Gauge Price Quantity  
SG000655 SGE™
SG000655
Removable 0.018 in. 2 in. 26 ga. Each for $254.54
Specifications
Description
  • Accuracy and reproducibility: ±1% (dispensed volume) (±2% for 0.5μL and 1μL syringes)
  • Borosilicate glass barrel outer diameter (OD): 0.5μL = 6.5mm, 1μL = 8mm, 5μL to 250μL = 6.5mm
  • Scale Length: 0.5μL = 27.05mm, 1μL to 250μL = 54.1mm
  • International Standards traceability
  • Needle Point Style: Cone
Description & Specifications
Specifications
GC Autosampler Syringe
Gas chromatography
Cone
Each