Please login to your online account to display your discounted pricing
SGE™ Syringes with Valves
Prefitted with removable needle and valve or Luer Lock valve
Manufacturer: sge™  SG008970
 View more versions of this product
Catalog No. SG-008970
  • / Each
Description & Specifications
Specifications
Material Borosilicate glass, PTFE plunger
Volume (Metric) 10mL
Autoclavable Yes
For Use With (Application) For sample storage and transportation or to pressurize sample prior to injection
Connection Type Removable
Termination Luer-lock
Type Syringe