Please login to your online account to display your discounted pricing
SGE™ Stylet Wires for Syringe Cleaning
Manufacturer: sge™  031746
 View more versions of this product
Catalog No. SG031746
  • / Pack of 5 Each
Description & Specifications
Specifications
Length (Metric) 15cm
Type Replacement Stylet Wire
For Use With (Equipment) 15 to 500μL syringe needles
Length (English) 5.9 in.