missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGE™ Split Straight-Through GC Inlet Liners

Supplied in blister packs to prevent contamination

$107.59 - $425.41

Specifications

Diameter (Metric) Inner 0.4cm
Diameter (Metric) Outer 0.63cm
For Use With (Equipment) Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
Length (Metric) 7.85cm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
SG092007 SGE™
SG092007
5/Pack Each for $107.59
SG092222 SGE™
SG092222
25/Pack Pack of 25 Each for $425.41
Specifications

Description

  • For various GC instruments
  • All liners are deactivated using high temperature (420°C) gas phase deactivation
  • Reduced active sites mean no unwanted reaction in samples

Description & Specifications

Specifications

0.4cm
0.63cm
Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
7.85cm