Please login to your online account to display your discounted pricing

SGE™ Split Straight-Through GC Inlet Liners


Supplied in blister packs to prevent contamination

Manufacturer: sge™  SG092222

 View more versions of this product

Catalog No. SG092222

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Product Type GC split straight-through liner
Diameter (Metric) Inner 0.4cm
For Use With (Equipment) Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
Quantity 25/Pack
Length (Metric) 7.85cm
Diameter (Metric) Outer 0.63cm