Please login to your online account to display your discounted pricing
SGE™ SilTite™ FingerTite Ferrule System Starter Kits
For gas chromatography systems
Manufacturer: sge™  SG073612
 View more versions of this product
Catalog No. SG073612
  • / Each
Description & Specifications
Specifications
Type Fingertite Shimadzu Agilent INJ/MS Starter Kit
For Use With (Equipment) 0.53mm I.D. columns