Please login to your online account to display your discounted pricing
SGE™ SilTite™ Ferrule Starter Kits
Specially designed ferrule and nut system developed specifically for fused silica GC columns and GC/MS systems
Manufacturer: sge™  SG073200
 View more versions of this product
Catalog No. SG073200
  • / Each
Description & Specifications
Specifications
Product Type Ferrule kit
Quantity 10/Pack
For Use With (Equipment) 0.1 to 0.25mm I.D. Column
Includes Ten 0.4mm I.D. ferrules and 2 nuts
For Use With (Application) GC/MS interface connections
Material Metal