Please login to your online account to display your discounted pricing
SGE™ SilTite™ Ferrule Starter Kits Replacement SilTite Nuts and Base Seals
Specially designed ferrule and nut system developed specifically for fused silica GC columns and GC/MS systems
Manufacturer: sge™  SG073224
 View more versions of this product
Catalog No. SG073224
  • / Pack of 5 Each
Description & Specifications
Specifications
Quantity 5/Pack
Product Type Metal Nut
Material Metal
For Use With (Equipment) SilTite™ Metal ferrule
For Use With (Application) Split/splitless injector