SGE™ SilTite™ Ferrule Starter Kits Replacement SilTite Nuts and Base Seals
Specially designed ferrule and nut system developed specifically for fused silica GC columns and GC/MS systems
$114.74 - $531.09
Specifications
For Use With (Application) Split/splitless injector
For Use With (Equipment) SilTite™ Metal ferrule
Material Metal
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
SG073224 SGE™
SG073224
Metal Nut 5/Pack Pack of 5 Each for $114.74
SG073226 SGE™
SG073226
Metal Nut 5/Pack Pack of 5 Each for $156.13
SG073401 SGE™
SG073401
Inlet Gold base seal 10/Pack Pack of 10 Each for $531.09
Specifications
Description
For use with SGE SilTite Ferrule Starter Kits
SilTite Nuts
  • Non-Swagelok™ nuts with two notches on outer surface
  • Inner taper of the nut corresponds to angle of ferrule, ensuring an even closure of the ferrule around the whole column to eliminate leaks
  • Ferrules are easily removed from nut while still attached to capillary column
  • Reusable
Description & Specifications
Specifications
Split/splitless injector
SilTite™ Metal ferrule
Metal