Please login to your online account to display your discounted pricing

SGE™ SilTite™ Ferrule Starter Kits Replacement Ferrule

Specially designed ferrule and nut system developed specifically for fused silica GC columns and GC/MS systems

$88.28 - $88.80

Specifications

For Use With (Application) All connections
Material Metal
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inside For Use With (Equipment) Product Type Quantity Price Quantity    

SG073220

 
sge™
SG073220
0.4mm 0.1 to 0.25mm I.D. Column Ferrule 10/Pack Pack of 10 Each for $88.28

SG073222

 
sge™
SG073222
0.8mm 0.53mm I.D. Column Ferrule 10/Pack Pack of 10 Each for $88.80

SG073221

 
sge™
SG073221
0.5mm 0.32mm I.D. Column Ferrule 10/Pack Pack of 10 Each for $88.28
Description & Specifications

Specifications

For Use With (Application) All connections
Material Metal

For use with SGE SilTite Ferrule Starter Kits

  • Unique 100% noncontaminating SilTite metal does not need retightening after use
  • Form a strong permanent airtight seal around the capillary column
  • Ferrule base is flat to form tight leak-resistant seal with MS interface
  • Ideal for high pressure applications
  • Maximum temperature: 500°C (932°F)
  • Must be used with supplied SilTite nut