Please login to your online account to display your discounted pricing
SGE™ SilFlow™ GC 4 Port Splitter
Highly efficient and reliable micro fluidic platform to improve GC connectivity
Manufacturer: sge™  123731
 View more versions of this product
Catalog No. 03-350-238
  • / Each
Description & Specifications
Specifications
Diameter (Metric) Outer N/A
Includes SilFlow microchannel device (MCD), SilTite FingerTite tool, Mounting bracket, Appropriately sized ferrules, nuts and blanking ferrules for set up
For Use With (Equipment) SGE Silflow microchannel device
Diameter (Metric) Inner 0.53mm ID
Type Port Splitter