Please login to your online account to display your discounted pricing
SGE™ SilFlow™ GC 3 Port Splitter Kits
Highly efficient and reliable micro fluidic platform to improve GC connectivity.
Manufacturer: sge™  123721
 View more versions of this product
Catalog No. 03-350-232
  • / Each
Description & Specifications
Specifications
Includes SilFlow microchannel device (MCD), SilTite FingerTite tool, Mounting bracket, Appropriately sized ferrules, nuts and blanking ferrules for set up
Diameter (Metric) Inner 0.53mm ID
Type Port Splitter
For Use With (Equipment) SGE Silflow microchannel device