Please login to your online account to display your discounted pricing
SGE™ SilFlow™ GC 3 Port Splitter: Device Only
Highly efficient and reliable micro fluidic platform to improve GC connectivity
Manufacturer: sge™  123724
 View more versions of this product
Catalog No. 03-350-235
  • / Each
Description & Specifications
Specifications
For Use With (Equipment) SGE Silflow Microchannel Device
Type Port Splitter
Diameter (Metric) Inner 0.53mm OD