Please login to your online account to display your discounted pricing
SGE™ SilFlow™ GC 3 Port Splitter: Device Only
Highly efficient and reliable micro fluidic platform to improve GC connectivity
$1,037.96 - $1,037.96
Specifications
Type Port Splitter
For Use With (Equipment) SGE Silflow Microchannel Device
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity    
03-350-234
sge™
123723
1.1mm OD Each for $1,037.96
03-350-235
sge™
123724
0.53mm OD Each for $1,037.96
03-350-236
sge™
123725
0.25/0.32mm ID Each for $1,037.96
Description & Specifications
Specifications
Type Port Splitter
For Use With (Equipment) SGE Silflow Microchannel Device
  • 1.1mm OD tubing not included