Please login to your online account to display your discounted pricing
SGE™ Replacement Plungers
Manufacturer: sge™  SG0318120
 View more versions of this product
Catalog No. SG0318120
  • / Pack of 2 Each
Description & Specifications
Specifications
For Use With Syringes 002987, 002991
Type Replacement Plunger, P10F-C/T-GT
For Use With (Application) IN VITRO use only. For lab analyzers only. Not for human use.
For Use With (Equipment) For 10uL fixed needle gas tight syringe for CTC/Carlo Erba/ Fisons autosampler
Material PTFE tip