Please login to your online account to display your discounted pricing
SGE™ Replacement Plungers
Manufacturer: sge™  SG031811
 View more versions of this product
Catalog No. SG031811
  • / Pack of 2 Each
Description & Specifications
Specifications
For Use With Syringes 002250, 002500, 002313
Type Replacement Plunger, P10R-GT
For Use With (Application) IN VITRO use only. For lab analyzers only. Not for human use.
For Use With (Equipment) For10uL removable needle gas tight syringe
Material PTFE tip