SGE™ Replacement Plungers
Manufacturer:  SGE™ SG031218
 View more versions of this product
Catalog No. SG031218
Specifications Description & Specifications
Specifications
IN VITRO use only. For lab analyzers only. Not for human use.
For 10uL Varian autosampler syringes; Syringes 002926, 002924, 002923
PTFE tip
Replacement Plunger, 10A-VP