SGE™ Replacement Needles for 5uL eVol™ NMR Syringes
For 5uL eVol NMR Syringes
$65.07 - $65.07
Specifications
Diameter (Metric) Outer 0.5mm
For Use With (Equipment) 5μL eVol™ NMR syringes
Needle Gauge 25 ga.
Material Stainless steel
Needle Point Style Bevel
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Needle Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Length (Metric) Needle Price Quantity  
SG036910 SGE™
SG036910
0.11mm 50mm Pack of 5 Each for $65.07
Specifications Description & Specifications
Specifications
0.5mm
25 ga.
Bevel
5μL eVol™ NMR syringes
Stainless steel
Replacement Needle