SGE™ Replacement Needles for 10μL Syringes
Made of Stainless steel
Manufacturer:  SGE™ SG037110
 View more versions of this product
Catalog No. SG-037110
Specifications
Description
Needle, Replacement; SGE; For 10μL syringe; Bevel point style; Stainless steel; Removable; 50mm L; 26 gauge
Made of Stainless steel
Description & Specifications
Specifications
0.004 in.
0.11mm
For 10μL syringes
51mm
Bevel
26 ga.
0.01 in.
0.25mm
2 in.
Stainless steel
Replacement Needle