missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGE™ Replacement Needles for 1 - 2.5mL Syringes

Stainless steel

Manufacturer:  SGE™ SG039060

 View more versions of this product

Catalog No. 03-350-260


 

Description

Description

  • 0.63mm OD, 0.32mm ID
  • 23 gauge

Specifications

Specifications

0.012 in.
0.02 in.
0.5mm
4.52 in.
115mm
Replacement Needle
23 ga.
0.32mm
For 1 to 2.5μL syringes
Stainless steel
Cone
Documents