SGE™ Replaceable Luer Lock Needles: Bevel
Designed for optimum septum penetration and prevention of septum coring.
$85.54 - $85.54
Specifications
For Use With (Application) Gas chromatography
Material Stainless Steel
Needle Point Style Bevel
Termination Luer-lock
Type Replacement Needle
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Gauge Length (Metric) Needle Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Gauge Length (Metric) Needle Price Quantity  
SG-039811 SGE™
SG039811
0.32mm 0.63mm 23 ga 115mm Pack of 5 Each for $85.54
Specifications
Description
  • Standard general purpose needle
  • Made of stainless steel
  • Preferred for manual injection where piercing the septum in exactly the same place is difficult
Description & Specifications
Specifications
Gas chromatography
Bevel
Replacement Needle
Stainless Steel
Luer-lock