Please login to your online account to display your discounted pricing
SGE™ GC Septa: CS
Designed specifically for use in Agilent Technologies™ (HP) GC instruments
Manufacturer: sge™  SG041828
 View more versions of this product
Catalog No. SG041828
  • / Pack of 50 Each
Description & Specifications
Specifications
Diameter (English) Inner 0.5 in.
Material CS material (PTFE-Coated Silicone)
Packaging Quantity 50/Pk.