Please login to your online account to display your discounted pricing
SGE™ GC Septa: Auto-Sep T
Designed specifically for use in Agilent Technologies™ (HP) GC instruments
$289.26 - $289.26
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    
SG041883
sge™
SG041883
100/Pk. Pack of 100 Each for $289.26
Description & Specifications
Auto-Sep T Septa
  • PTFE-coated silicone septa for temperature program injectors and high-volume autosamplers (especially for GC/MS)
  • Excellent durability, resealing, solvent and tear resistance
  • Maximum temperature: 350°C (662°F)