Please login to your online account to display your discounted pricing
SGE™ GC Inlet Tapered FocusLiners™ Split/Splitless
Results in lower mass discrimination
Manufacturer: sge™  SG092003
 View more versions of this product
Catalog No. SG092003
  • / Pack of 5 Each
Description & Specifications
Specifications
Product Type Split/splitless tapered focus liner
Diameter (Metric) Inner 0.4cm
Diameter (Metric) Outer 0.63cm
Length (Metric) 7.85cm
Quantity 5/Pack
For Use With (Equipment) Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177