Please login to your online account to display your discounted pricing

SGE™ GC Inlet FocusLiners™


Results in lower mass discrimination

Manufacturer: sge™  SG092219

 View more versions of this product

Catalog No. SG092219

  • / Pack of 25 Each
 

Description & Specifications

Specifications

Product Type Split/splitless fast focus liner
Packing Material Quartz Wool
Length (Metric) 7.85cm
For Use With (Equipment) Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
Quantity 25/Pack
Diameter (Metric) Inner 0.4cm
Diameter (Metric) Outer 0.63cm