Please login to your online account to display your discounted pricing

SGE™ GC Inlet FocusLiners™


Results in lower mass discrimination

Manufacturer: sge™  SG092002

 View more versions of this product

Catalog No. SG092002

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Product Type Split/splitless fast focus liner
Packing Material Quartz Wool
Length (Metric) 7.85cm
Quantity 5/Pack
For Use With (Equipment) Agilent HP 5890, 6890, 7890, 6850, 4890, Thermo 9001, GCQ TRACE 1300, Varian 1177
Diameter (Metric) Outer 0.63cm
Diameter (Metric) Inner 0.4cm