Please login to your online account to display your discounted pricing
SGE™ SilTite™ GC Capillary Column Connectors: μ Unions
Manufacturer: sge™  SG073560
 View more versions of this product
Catalog No. SG073560
  • / Each
Description & Specifications
Specifications
Includes 2 unions, 5 double taper ferrules and tools
For Use With GC Capillary Column
Connections 0.1-0.25mm and 0.1-0.25mm
Type Connector