Please login to your online account to display your discounted pricing
SGE™ GC Capillary Column Connectors: Replacement Ferrules
Manufacturer: sge™  SG073569
 View more versions of this product
Catalog No. SG073569
  • / Pack of 10 Each
Description & Specifications
Specifications
Type Replacement SilTite™ μ-Union ferrules
Material Stainless Steel
For Use With 0.32mm I.D. First column and second column with 0.5mm holes