Please login to your online account to display your discounted pricing
SGE™ GC Capillary Column Connectors: Replacement Ferrules
Manufacturer: sge™  SG073567
 View more versions of this product
Catalog No. SG073567
  • / Pack of 10 Each
Description & Specifications
Specifications
Material Stainless Steel
Type Replacement SilTite™ μ-Union ferrules
For Use With 0.1 to 0.25mm I.D. first column with 0.4mm holes and 0.32mm I.D. second column with 0.5mm holes