Please login to your online account to display your discounted pricing
SGE™ Gastight Syringes: Fixed Needle Models
Suitable for liquid and gas samples
Manufacturer: sge™  SG005200
 View more versions of this product
Catalog No. SG-005200
  • / Each
Description & Specifications
Specifications
Capacity (Metric) 100μL
Material (Tip) PTFE
For Use With (Application) Suitable for gases or liquids for maximum inertness
Gauge 25 ga
Precision ±1%
Material (Barrel) Borosilicate Glass
Model 25F-GT