missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SGE™ Gas Tight Syringe with Removable Needle

Ideal for sample storage, transportation and pressurizing samples prior to injection

$127.28 - $127.28

Specifications

Connection Type Removable
Diameter (English) Outer Needle 0.024 in.
Diameter (Metric) Outer Needle 0.63mm
Type Analytical Syringe
Needle Gauge 23 ga.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Length (English) Needle Model Needle Point Style Price Quantity  
Catalog Number Mfr. No. Length (English) Length (English) Needle Model Needle Point Style Price Quantity  
SG002902 SGE™
002902
1.65 in. 1.65 in. 10R-S-GT-0.63C Removable Cone Each for $127.28
Specifications

Description

  • Removable needle
  • Gas tight plunger
  • Stainless steel
  • Gauge: 23

Description & Specifications

Specifications

Removable
0.63mm
23 ga.
0.024 in.
Analytical Syringe
Each