SGE™ Gas Tight Syringe with Removable Needle
Ideal for sample storage, transportation and pressurizing samples prior to injection
Manufacturer:  SGE™ 002902
 View more versions of this product
Catalog No. SG002902
Specifications Description & Specifications
Specifications
Removable
0.63mm
1.65 in.
42mm
10R-S-GT-0.63C
Each
23 ga.
0.024 in.
Shimadzu AOC14, AOC17 and AOC20 autosampler
1.65 in.
42mm
Removable Cone
Analytical Syringe
10μL