missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SGE™ Gas Tight Plunger Syringe with PTFE Luer Lock
Headspace syringes with a special Luer lock probe
Manufacturer: SGE™ SG008975
Description
- For soil gas sampling
- Special Luer lock probe has a side hole for efficient sample intake
- Fitted with a valve (SVLL)
- Gas tight
- Volume: 10mL
Specifications
Plunger Syringe | |
PTFE luer-lock valve | |
10.0 mL |
For soil gas sampling | |
Each |