Please login to your online account to display your discounted pricing
SGE™ BPX35 GC Capillary Columns
$914.37 - $914.37
Specifications
Coating Fused Silica
Functionality/Interaction Type Pi Bonding with the Aromatic Phenyl Groups
Packing Material Forte™ BPX35/BPX608
Technique GC-MS
For Use With (Application) Conformational Analysis, Environmental Analyses, Pesticides/herbicides, Drugs of Abuse, Pharmaceuticals, Polynuclear Aromatic Hydrocarbons, GC-MS applications
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity    
SG054701
sge™
SG054701
0.25mm 0.25μm 30m Each for $914.37
Description & Specifications
Specifications
Coating Fused Silica
Functionality/Interaction Type Pi Bonding with the Aromatic Phenyl Groups
Packing Material Forte™ BPX35/BPX608
Technique GC-MS
For Use With (Application) Conformational Analysis, Environmental Analyses, Pesticides/herbicides, Drugs of Abuse, Pharmaceuticals, Polynuclear Aromatic Hydrocarbons, GC-MS applications
Max. Temperature 330/360°C
Polarity Mid Polarity
Stationary Phase 35% Phenyl Polysilphenylene-siloxane
USP Type G28, G32, G42