Please login to your online account to display your discounted pricing
SGE™ BP5 GC Capillary Columns
$1,143.54 - $1,143.54
Specifications
Coating Fused Silica
Packing Material Forte™ BPX5
For Use With (Application) For GC-MS, Ultra Trace Analyses, Pesticides/herbicides, Hydrocarbons, Solvents, Phenols, Amines, GC-MS and Other Specific Detector Applications
Max. Temperature 350/360°C
Polarity Non Polar
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity    
SG054160
sge™
SG054160
0.53mm 3μm 30m Each for $1,143.54
Description & Specifications
Specifications
Coating Fused Silica
Packing Material Forte™ BPX5
For Use With (Application) For GC-MS, Ultra Trace Analyses, Pesticides/herbicides, Hydrocarbons, Solvents, Phenols, Amines, GC-MS and Other Specific Detector Applications
Max. Temperature 350/360°C
Polarity Non Polar
Stationary Phase 5% Phenyl Polysilphenylene-siloxane
USP Type G27, G36, G41